4-Picolyl bromide hydrobromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-(Bromomethyl)pyridine hydrobromide

B3315-5G B3315-25G
TCB3315-5GEA 109 USD
TCB3315-5G TCB3315-25G
4-Picolyl bromide hydrobromide ≥97.0% (by titrimetric analysis)
4-Picolyl bromide hydrobromide

CAS Number: 73870-24-3
MDL Number: MFCD01863545
Molecular Formula: C6H6BrN
Molecular Weight: 252.94
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 186

Formula: C₆H₇Br₂N
MW: 252.94 g/mol
Melting Pt: 186 °C
MDL Number: MFCD01863545
CAS Number: 73870-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44b5662eaf73f4299a2cbcf9cacee2c0c50a6e40&vid=44b5662eaf73f4299a2cbcf9cacee2c0c50a6e40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR