(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene ≥92.0% (by GC)

Supplier: TCI America

B3298-1G B3298-5G
TCB3298-1GEA 173.7 USD
TCB3298-1G TCB3298-5G
(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene ≥92.0% (by GC)
(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene

CAS Number: 72658-10-7
Molecular Formula: C10H22O2Si
Molecular Weight: 202.37
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Boiling point (°C): 53
Storage Temperature: 0-10°C

Formula: C₁₀H₂₂O₂Si
MW: 202.37 g/mol
Boiling Pt: 53 °C
CAS Number: 72658-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ff2fc51e5b9da42227c481b38d572cd86d05bef&vid=1ff2fc51e5b9da42227c481b38d572cd86d05bef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >92.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR