4-Bromoimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3280-1G B3280-5G
TCB3280-1GEA 22 USD
TCB3280-1G TCB3280-5G
4-Bromoimidazole ≥98.0% (by GC, titration analysis)
4-Bromoimidazole

CAS Number: 2302-25-2
MDL Number: MFCD00047021
Molecular Formula: C3H3BrN2
Molecular Weight: 146.98
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 133

Formula: C₃H₃BrN₂
Melting Pt: 133 °C
MDL Number: MFCD00047021
CAS Number: 2302-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4efc13fc6dd4b7eb0655dce88adb932afe9506d8&vid=4efc13fc6dd4b7eb0655dce88adb932afe9506d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR