4-Bromo-3,3,4,4-tetrafluoro-1-butene ≥98.0% (by GC)

Supplier: TCI America

B3222-25G B3222-5G
TCB3222-25GEA 166 USD
TCB3222-25G TCB3222-5G
4-Bromo-3,3,4,4-tetrafluoro-1-butene ≥98.0% (by GC)
4-Bromo-3,3,4,4-tetrafluoro-1-butene

CAS Number: 18599-22-9
MDL Number: MFCD00042189
Molecular Formula: C4H3BrF4
Molecular Weight: 206.97
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Specific Gravity (20/20): 1.63

Formula: C₄H₃BrF₄
MW: 206.97 g/mol
Boiling Pt: 60 °C
Density: 1.63 (20 °C)
MDL Number: MFCD00042189
CAS Number: 18599-22-9
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dd6dd94a1b000b924b2dc8345bb3dcf0b8f2e1f&vid=48fde55598cae802f1a00bce5a50198d441512c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR