Bicalutamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulphonyl)-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulphonyl]-2-hydroxy-2-methylpropionamide

B3206-1G B3206-200MG
TCB3206-1GEA 203 USD
TCB3206-1G TCB3206-200MG
Bicalutamide ≥98.0% (by HPLC, total nitrogen)
Bicalutamide

CAS Number: 90357-06-5
MDL Number: MFCD00869971
Molecular Formula: C18H14F4N2O4S
Molecular Weight: 430.37
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 196
Lambda max.: 270 nm (CH3CN)


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Antitumor Ingredients
Other Product Names / Synonyms: N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropionamide

Formula: C₁₈H₁₄F₄N₂O₄S
MW: 430.37 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00869971
CAS Number: 90357-06-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a96f031ac0de90c2380cb11202ced270f8631010&vid=a96f031ac0de90c2380cb11202ced270f8631010&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR