2-Bromo-3-(trifluoromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

B3179-1G B3179-5G
TCB3179-1GEA 27 USD
TCB3179-1G TCB3179-5G
2-Bromo-3-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Bromo-3-(trifluoromethyl)pyridine

CAS Number: 175205-82-0 MDL Number: MFCD00153084 Molecular Formula: C6H3BrF3N Molecular Weight: 226.00 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 69 Melting point (°C): 40

Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 69 °C
Melting Pt: 40 °C
MDL Number: MFCD00153084
CAS Number: 175205-82-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8a52ac26f6c86d81a5e974a46d16d012337ff1e&vid=e8a52ac26f6c86d81a5e974a46d16d012337ff1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR