4-Bromo-2-iodoaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B3164-1G B3164-5G
TCB3164-1GEA 23 USD
TCB3164-1G TCB3164-5G
4-Bromo-2-iodoaniline ≥97.0% (by GC, titration analysis)
4-Bromo-2-iodoaniline

CAS Number: 66416-72-6
MDL Number: MFCD02093764
Molecular Formula: C6H5BrIN
Molecular Weight: 297.92
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 70

Formula: C₆H₅BrIN
MW: 297.92 g/mol
Melting Pt: 70 °C
MDL Number: MFCD02093764
CAS Number: 66416-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78cf5f3186260da00c85c735e6ff80dc11302d96&vid=78cf5f3186260da00c85c735e6ff80dc11302d96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR