3-(Bromomethyl)-5-chlorobenzo[b]thiophene ≥98.0% (by GC)

Supplier: TCI America

B3147-1G B3147-5G
TCB3147-1GEA 26 USD
TCB3147-1G TCB3147-5G
3-(Bromomethyl)-5-chlorobenzo[b]thiophene ≥98.0% (by GC)
3-(Bromomethyl)-5-chlorobenzo[b]thiophene

CAS Number: 1198-51-2
MDL Number: MFCD00173769
Molecular Formula: C9H6BrClS
Molecular Weight: 261.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 131
Storage Temperature: 0-10°C

Formula: C₉H₆BrClS
MW: 261.5700 g/mol
Melting Pt: 131 °C
MDL Number: MFCD00173769
CAS Number: 1198-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0fa2f402c92a3bcc97eebfc40a3f1b26029135c&vid=e0fa2f402c92a3bcc97eebfc40a3f1b26029135c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR