Benzyltriethylammonium borhydride ≥90.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Benzyltriethylammonium Borohydride

B3128-25G B3128-5G
TCB3128-25GEA 362 USD
TCB3128-25G TCB3128-5G
Benzyltriethylammonium borhydride ≥90.0% (by titrimetric analysis)
Benzyltriethylammonium borhydride

CAS Number: 85874-45-9
MDL Number: MFCD00191785
Molecular Formula: C13H26BN
Molecular Weight: 207.17
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Melting point (°C): 143

Formula: C₁₃H₂₆BN
MW: 207.17 g/mol
Melting Pt: 143 °C
MDL Number: MFCD00191785
CAS Number: 85874-45-9
UN: 3131
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=725a1556adcbe3b24cece810dba76f13b249da61&vid=725a1556adcbe3b24cece810dba76f13b249da61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR