4-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America

B3120-1G B3120-5G
TCB3120-1GEA 23 USD
TCB3120-1G TCB3120-5G
4-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)
4-Bromo-2-hydroxybenzonitrile

CAS Number: 288067-35-6
MDL Number: MFCD07780675
Molecular Formula: C7H4BrNO
Molecular Weight: 198.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: 5-Bromo-2-cyanophenol

Formula: C₇H₄BrNO
MW: 198.01 g/mol
Melting Pt: 160 °C
MDL Number: MFCD07780675
CAS Number: 288067-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca1531d92fe1e03fc1450d52770368eb1a66a639&vid=96ca1e016071df32fb118b94c8cf3b1263f3f1dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR