3-Bromo-2-(bromomethyl)propionic acid ≥98.0%
Supplier: TCI America
Synonyms:
beta,beta'-Dibromoisobutyric Acid, Bis(bromomethyl)acetic Acid, 3-Bromo-2-(bromomethyl)propanoic acid
B3081-25G
B3081-5G
TCB3081-25GEA
226
USD
TCB3081-25G
TCB3081-5G
3-Bromo-2-(bromomethyl)propionic acid ≥98.0%
3-Bromo-2-(bromomethyl)propionic acid
CAS Number: 41459-42-1
MDL Number: MFCD00010643
Molecular Formula: C4H6Br2O2
Molecular Weight: 245.90
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 99
Storage Temperature: 0-10°C
Other Product Names / Synonyms: beta,beta'-Dibromoisobutyric Acid, Bis(bromomethyl)acetic Acid
Formula:
C₄H₆Br₂O₂ Melting Pt: 99 °C |
MDL Number:
MFCD00010643 CAS Number: 41459-42-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...