4-Bromobenzyl chloride ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene

B3042-25G B3042-5G
TCB3042-25GEA 50 USD
TCB3042-25G TCB3042-5G
4-Bromobenzyl chloride ≥98.0%
4-Bromobenzyl chloride

CAS Number: 589-17-3
MDL Number: MFCD00040867
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 236
Melting point (°C): 39


Other Product Names / Synonyms: 4-Bromo-alpha-chlorotoluene

Formula: C₇H₆BrCl
Boiling Pt: 236 °C
Melting Pt: 39 °C
MDL Number: MFCD00040867
CAS Number: 589-17-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb78fd952a4bd26213b2bcf6d3e055d0ee066d22&vid=e667da66176a2e49a4a74a7dfb02681ab36b950f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR