Borane-2-methylpyridine complex ≥85.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Picoline Borane, 2-Methylpyridine Borane, Borane - 2-Picoline Complex

B3018-25G B3018-5G
TCB3018-25GEA 216.6 USD
TCB3018-25G TCB3018-5G
Borane-2-methylpyridine complex ≥85.0% (by titrimetric analysis)
Borane-2-methylpyridine complex

CAS Number: 3999-38-0
MDL Number: MFCD07784361
Molecular Formula: C6H10BN
Molecular Weight: 106.96
Purity/Analysis Method: >85.0% (T)
Form: Crystal
Melting point (°C): 48
Flash Point (°C): 100


Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Borane - 2-Picoline Complex, 2-Picoline Borane, 2-Methylpyridine Borane

Formula: C₆H₁₀BN
MW: 106.96 g/mol
Melting Pt: 48 °C
Flash Pt: 100 °C
MDL Number: MFCD07784361
CAS Number: 3999-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fbb7a25383e32ac4333432fc28328e87d4b52e3&vid=1fbb7a25383e32ac4333432fc28328e87d4b52e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR