4-Butylbenzyl bromide ≥97.0% (by GC)

Supplier: TCI America

B3005-1G B3005-5G
TCB3005-1GEA 91 USD
TCB3005-1G TCB3005-5G
4-Butylbenzyl bromide ≥97.0% (by GC)
4-Butylbenzyl bromide

CAS Number: 10531-16-5
Molecular Formula: C11H15Br
Molecular Weight: 227.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Specific Gravity (20/20): 1.22

Formula: C₁₁H₁₅Br
MW: 227.15 g/mol
Boiling Pt: 144 °C
Density: 1.22 (20 °C)
CAS Number: 10531-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33b6611aeed0b4608293bb13b6e7287a534dade2&vid=33b6611aeed0b4608293bb13b6e7287a534dade2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR