α,3-Dibromo-4-fluorotoluene ≥97.0%

Supplier: TCI America

Synonyms: 3-Bromo-4-fluorobenzyl bromide, 2-Bromo-4-(bromomethyl)-1-fluorobenzene

B2975-25G B2975-5G
TCB2975-25GEA 163.2 USD
TCB2975-25G TCB2975-5G
α,3-Dibromo-4-fluorotoluene ≥97.0%
α,3-Dibromo-4-fluorotoluene

CAS Number: 78239-71-1
Molecular Formula: C7H5Br2F
Molecular Weight: 267.92
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 41

Formula: C₇H₅Br₂F
Melting Pt: 41 °C
MDL Number: MFCD09037367
CAS Number: 78239-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49cbcc71c562720f7d9ba77459e56af02dc5667b&vid=49cbcc71c562720f7d9ba77459e56af02dc5667b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR