1-Bromo-2,3,5-trifluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,3,5-Trifluorobromobenzene

B2970-1G B2970-5G
TCB2970-1GEA 27 USD
TCB2970-1G TCB2970-5G
1-Bromo-2,3,5-trifluorobenzene ≥98.0% (by GC)
1-Bromo-2,3,5-trifluorobenzene

CAS Number: 133739-70-5
MDL Number: MFCD00012232
Molecular Formula: C6H2BrF3
Molecular Weight: 210.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 32
Specific Gravity (20/20): 1.79

Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Density: 1.79 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00012232
CAS Number: 133739-70-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05550599e4afb3906a4d8a0f6966d9ff25e72d1a&vid=05550599e4afb3906a4d8a0f6966d9ff25e72d1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR