2-sec-Butyl-3-methoxypyrazine ≥97.0% (by GC)

Supplier: TCI America

B2959-1G B2959-5G
TCB2959-1GEA 47 USD
TCB2959-1G TCB2959-5G
2-sec-Butyl-3-methoxypyrazine ≥97.0% (by GC)
2-sec-Butyl-3-methoxypyrazine

CAS Number: 24168-70-5
MDL Number: MFCD00006136
Molecular Formula: C9H14N2O
Molecular Weight: 166.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 77
Specific Gravity (20/20): 1.00

Formula: C₉H₁₄N₂O
Boiling Pt: 99 °C
Density: 1.00 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00006136
CAS Number: 24168-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c2a3513b80449fab9aca3b9a73ab4651a617cc8&vid=2c2a3513b80449fab9aca3b9a73ab4651a617cc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR