1,6-Bis(acryloyloxy)hexane ≥85.0% stabilised

Supplier: TCI America

Synonyms: HDODA, Hexamethylene Glycol Diacrylate, Hexamethylene diacrylate, 1,6-Hexanediol Diacrylate

B2936-25G B2936-500G
TCB2936-25GEA 28.2 USD
TCB2936-25G TCB2936-500G
1,6-Bis(acryloyloxy)hexane ≥85.0% stabilised
1,6-Bis(acryloyloxy)hexane

(stabilized with MEHQ)
CAS Number: 13048-33-4
MDL Number: MFCD00008631
Molecular Formula: C12H18O4
Molecular Weight: 226.27
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 113
Specific Gravity (20/20): 1.02


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,6-Hexanediol Diacrylate, Hexamethylene Glycol Diacrylate, HDODA

Formula: C₁₂H₁₈O₄
Boiling Pt: 165 °C
Density: 1.02 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008631
CAS Number: 13048-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=511db893fe167bd7600154933f941fb9618e7c25&vid=511db893fe167bd7600154933f941fb9618e7c25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR