1,4-Butanediol diacrylate ≥90.0% stabilised

Supplier: TCI America

B2935-25G B2935-500G
TCB2935-25GEA 44.49 USD
TCB2935-25G TCB2935-500G
1,4-Butanediol diacrylate ≥90.0% stabilised
1,4-Butanediol diacrylate

(stabilized with MEHQ)
CAS Number: 1070-70-8
MDL Number: MFCD00014940
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 125
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Butanediol Diacrylate, 1,4-Butylene Glycol Diacrylate, Tetramethylene Glycol Diacrylate

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Boiling Pt: 138 °C
Density: 1.05 (20 °C)
Flash Pt: 125 °C
MDL Number: MFCD00014940
CAS Number: 1070-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2d00eaf166daf35d4d8dd2c60c746df70aaedc8&vid=a2d00eaf166daf35d4d8dd2c60c746df70aaedc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR