(S)-(-)-3-Butyne-2-ol ≥98.0%
Supplier: TCI America
Synonyms:
(S)-3-Butyn-2-ol, (S)-(-)-3-Hydroxy-1-butyne, (S)-(-)-But-3-yn-2-ol
B2910-1G
B2910-5G
TCB2910-1GEA
128.32
USD
TCB2910-1G
TCB2910-5G
(S)-(-)-3-Butyne-2-ol ≥98.0%
(S)-(-)-3-Butyne-2-ol
CAS Number: 2914-69-4
MDL Number: MFCD00190166
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 26
Specific Gravity (20/20): 0.89
Specific rotation [a]20/D: -47 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: (S)-(-)-1-Butyn-3-ol
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 100 °C Density: 0.89 (20 °C) Flash Pt: 26 °C |
MDL Number:
MFCD00190166 CAS Number: 2914-69-4 UN: 2929 ADR: 6.1 II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...