(R)-(+)-3-Butyne-2-ol ≥98.0%

Supplier: TCI America

Synonyms: (R)-(+)-But-3-yn-2-ol, (R)-But-3-yn-2-ol, (R)-(+)-3-Hydroxy-1-butyne

B2909-1G B2909-5G
TCB2909-1GEA 81 USD
TCB2909-1G TCB2909-5G
(R)-(+)-3-Butyne-2-ol ≥98.0%
(R)-(+)-3-Butyne-2-ol

CAS Number: 42969-65-3
MDL Number: MFCD00211237
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 26
Specific Gravity (20/20): 0.89
Specific rotation [a]20/D: 47 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: (R)-(+)-1-Butyn-3-ol

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 103
Density: 0.89 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00211237
CAS Number: 42969-65-3
UN: 2929
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41d3132b98ee0a3fa8bcd5f733f4d7a4893bdf26&vid=41d3132b98ee0a3fa8bcd5f733f4d7a4893bdf26&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR