tert-Butyldiphenylsilyl trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: tert-Butyldiphenylsilyl Triflate, Trifluoromethanesulphonic acid tert-Butyldiphenylsilyl Ester

B2898-1G B2898-5G
TCB2898-1GEA 131 USD
TCB2898-1G TCB2898-5G
tert-Butyldiphenylsilyl trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
tert-Butyldiphenylsilyl trifluoromethanesulfonate

CAS Number: 92886-86-7
Molecular Formula: C17H19F3O3SSi
Molecular Weight: 388.48
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: tert-Butyldiphenylsilyl Triflate, Trifluoromethanesulfonic Acid tert-Butyldiphenylsilyl Ester

Formula: C₁₇H₁₉F₃O₃SSi
MW: 388.48 g/mol
MDL Number: MFCD01941212
CAS Number: 92886-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed2a2e8be311e20c0dd103e959151260ec68bc8d&vid=ed2a2e8be311e20c0dd103e959151260ec68bc8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR