Bis(trimethylsilyl)bromomethane ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Bis(trimethylsilyl)methyl Bromide

B2874-1G B2874-5G
TCB2874-1GEA 135 USD
TCB2874-1G TCB2874-5G
Bis(trimethylsilyl)bromomethane ≥95.0% (by GC)
Bis(trimethylsilyl)bromomethane

CAS Number: 29955-12-2
Molecular Formula: C7H19BrSi2
Molecular Weight: 239.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.09
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Bis(trimethylsilyl)methyl Bromide

Formula: C₇H₁₉BrSi₂
MW: 239.30 g/mol
Density: 1.09 (20 °C)
CAS Number: 29955-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9591d0d982ccaf90229ab817919d42d51443d8dd&vid=9591d0d982ccaf90229ab817919d42d51443d8dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR