5-Bromo-2,4-dichloropyrimidine ≥97.0% (by GC)

Supplier: TCI America

B2823-25G B2823-5G
TCB2823-25GEA 75 USD
TCB2823-25G TCB2823-5G
5-Bromo-2,4-dichloropyrimidine ≥97.0% (by GC)
5-Bromo-2,4-dichloropyrimidine

CAS Number: 36082-50-5
Molecular Formula: C4HBrCl2N2
Molecular Weight: 227.87
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 128
Flash Point (°C): 113
Freezing point (°C): 26

Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
Boiling Pt: 128 °C
Flash Pt: 113 °C
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c391db09ff380877552ce52f6e7579b2980f840&vid=3c391db09ff380877552ce52f6e7579b2980f840&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR