Bis(2,6-diisopropylphenyl)carbodiimide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

B2756-25G B2756-5G
TCB2756-25GEA 173 USD
TCB2756-25G TCB2756-5G
Bis(2,6-diisopropylphenyl)carbodiimide ≥98.0% (by total nitrogen basis)
Bis(2,6-diisopropylphenyl)carbodiimide

CAS Number: 2162-74-5
Molecular Formula: C25H34N2
Molecular Weight: 362.56
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 197

Formula: C₂₅H₃₄N₂
Melting Pt: 51 °C
Flash Pt: 197 °C
MDL Number: MFCD00082211
CAS Number: 2162-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ab4a351c93a3ac3f3816718abba47bcad68fe92&vid=1ab4a351c93a3ac3f3816718abba47bcad68fe92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR