2-Bromo-4-fluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

B2744-25G B2744-5G
TCB2744-25GEA 77 USD
TCB2744-25G TCB2744-5G
2-Bromo-4-fluorobenzonitrile ≥98.0% (by GC)
2-Bromo-4-fluorobenzonitrile

CAS Number: 36282-26-5
Molecular Formula: C7H3BrFN
Molecular Weight: 200.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77

Formula: C₇H₃BrFN
Melting Pt: 77 °C
MDL Number: MFCD00672924
CAS Number: 36282-26-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=900429c4afbad44e8656de295e95b11665fb7128&vid=6723b28423286320e082488d8bc9c3399218a65a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR