Bromine-1,4-dioxane complex ≥95.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 15481-39-7
Molecular Formula: C4H8Br2O2
Molecular Weight: 247.91
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 68
Storage Temperature: <0°C
Other Product Names / Synonyms: 1,4-Dioxane - Bromine Complex
Formula:
C₄H₈Br₂O₂ Boiling Pt: 95 °C Melting Pt: 68 °C |
MDL Number:
MFCD00034818 CAS Number: 15481-39-7 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...