2-Bromobenzenesulphonyl chloride ≥97.0%
Supplier: TCI America
Synonyms:
2-Bromobenzene-1-sulphonyl chloride
B2702-25G
B2702-5G
TCB2702-25GEA
205
USD
TCB2702-25G
TCB2702-5G
2-Bromobenzenesulphonyl chloride ≥97.0%
2-Bromobenzenesulphonyl chloride
CAS Number: 2905-25-1
Molecular Formula: C6H4BrClO2S
Molecular Weight: 255.51
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 117
Melting point (°C): 51
Formula:
C₆H₄BrClO₂S MW: 255.52 Boiling Pt: 117 °C Melting Pt: 51 |
MDL Number:
MFCD00051971 CAS Number: 2905-25-1 UN: 3261 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...