Benzoic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
B2635-25G
B2635-500G
TCB2635-25GEA
20.66
USD
TCB2635-25G
TCB2635-500G
Benzoic acid ≥99.0% (by GC, titration analysis)
Benzoic acid
CAS Number: 65-85-0
MDL Number: MFCD00002398
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method:
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 249 °C Melting Pt: 124 °C Flash Pt: 121 °C |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...