2-Bromo-4'-(methylsulfonyl)acetophenone ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Bromo-1-(4-(methylsulphonyl)phenyl)ethanone, 4-(Methylsulphonyl)phenacyl bromide
B2573-25G
B2573-5G
TCB2573-25GEA
146
USD
TCB2573-25G
TCB2573-5G
2-Bromo-4'-(methylsulfonyl)acetophenone ≥98.0% (by GC)
2-Bromo-4'-(methylsulphonyl)acetophenone
CAS Number: 50413-24-6
MDL Number: MFCD00673134
Molecular Formula: C9H9BrO3S
Molecular Weight: 277.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129
Formula:
C₉H₉BrO₃S MW: 277.1350 g/mol Melting Pt: 129 °C |
MDL Number:
MFCD00673134 CAS Number: 50413-24-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...