3-Bromoisobutyric acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Captopril EP Impurity D, 3-Bromo-2-methylpropanoic acid, 3-Bromo-2-methylpropionic Acid
B2571-25G
B2571-5G
TCB2571-25GEA
216.6
USD
TCB2571-25G
TCB2571-5G
3-Bromoisobutyric acid ≥97.0% (by GC, titration analysis)
3-Bromoisobutyric acid
CAS Number: 56970-78-6
Molecular Formula: C4H7BrO2
Molecular Weight: 167.00
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 231
Specific Gravity (20/20): 1.59
Other Product Names / Synonyms: 3-Bromo-2-methylpropionic Acid
Formula:
C₄H₇BrO₂ MW: 167.00 g/mol Boiling Pt: 231 °C Density: 1.59 (20 °C) |
MDL Number:
MFCD01310530 CAS Number: 56970-78-6 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...