1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B2475-25G B2475-5G
TCB2475-25GEA 235 USD
TCB2475-25G TCB2475-5G
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate

CAS Number: 402846-78-0
Molecular Formula: C9H17BF4N2
Molecular Weight: 240.05
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Freezing point (°C): 34

Formula: C₉H₁₇BF₄N₂
MDL Number: MFCD03427618
CAS Number: 402846-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce41a0ee8f34290a911bd6447b75790beb33d64a&vid=ce41a0ee8f34290a911bd6447b75790beb33d64a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR