5-Bromo-2-fluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

B2425-25G B2425-5G
TCB2425-25GEA 137 USD
TCB2425-25G TCB2425-5G
5-Bromo-2-fluorobenzonitrile ≥98.0% (by GC)
5-Bromo-2-fluorobenzonitrile

CAS Number: 179897-89-3
Molecular Formula: C7H3BrFN
Molecular Weight: 200.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Melting Pt: 80 °C
MDL Number: MFCD00143424
CAS Number: 179897-89-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f7bfe139e5505e39b43af0806e1509ba344cb04&vid=3f7bfe139e5505e39b43af0806e1509ba344cb04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR