sec-Butyl ethyl ether ≥97.0%
Supplier: TCI America
B2399-250G
B2399-25G
TCB2399-250GEA
834.8
USD
TCB2399-250G
TCB2399-25G
sec-Butyl ethyl ether ≥97.0%
sec-Butyl ethyl ether
CAS Number: 2679-87-0
MDL Number: MFCD00053773
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method:
Formula:
C₆H₁₄O Boiling Pt: 82 °C Density: 0.74 (20 °C) |
MDL Number:
MFCD00053773 CAS Number: 2679-87-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...