3-Bromobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2389-1G B2389-5G
TCB2389-1GEA 41.8 USD
TCB2389-1G TCB2389-5G
3-Bromobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3-Bromobenzenesulphonyl chloride

CAS Number: 2905-24-0
MDL Number: MFCD00052313
Molecular Formula: C6H4BrClO2S
Molecular Weight: 255.51
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.77

Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Density: 1.77 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00052313
CAS Number: 2905-24-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f1f4be3c7e492ce25e7b1ed97963026c797babb&vid=7f1f4be3c7e492ce25e7b1ed97963026c797babb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR