ω-Bromo-4-fluoroacetophenone ≥97.0%

Supplier: TCI America

Synonyms: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone

B2382-25G B2382-5G
TCB2382-25GEA 142.46 USD
TCB2382-25G TCB2382-5G
ω-Bromo-4-fluoroacetophenone ≥97.0%
ω-Bromo-4-fluoroacetophenone

CAS Number: 403-29-2
MDL Number: MFCD00040830
Molecular Formula: C8H6BrFO
Molecular Weight: 217.04
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 153
Melting point (°C): 49
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluorophenacyl Bromide

Formula: C₈H₆BrFO
Boiling Pt: 153 °C
Melting Pt: 49 °C
Flash Pt: 113 °C
MDL Number: MFCD00040830
CAS Number: 403-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9022b0bf59d334523002ed0a3e57c7d474275f84&vid=9022b0bf59d334523002ed0a3e57c7d474275f84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR