N,N-Bis(2-isopropoxyethyl)amine ≥97.0%

Supplier: TCI America
Danger

Synonyms: Bis(2-isopropoxyethyl)amine

B2356-25G B2356-5G
TCB2356-25GEA 411.1 USD
TCB2356-25G TCB2356-5G
N,N-Bis(2-isopropoxyethyl)amine ≥97.0%
N,N-Bis(2-isopropoxyethyl)amine

MDL Number: MFCD06797066 Molecular Formula: C10H23NO2 Molecular Weight: 189.30 Purity/Analysis Method: 97.0% (GC,T) Form: Clear Liquid Specific Gravity (20/20): 0.86

Formula: C₁₀H₂₃NO₂
Density: 0.86 (20 °C)
MDL Number: MFCD06797066
CAS Number: 1119451-05-6
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a0b271fc264907ffe8fc067696f0cb17c1d81d0&vid=6a0b271fc264907ffe8fc067696f0cb17c1d81d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR