3-Bromo-p-toluidine ≥98.0%
Supplier: TCI America
Synonyms:
4-Amino-2-bromotoluene, 3-Bromo-4-methylaniline
B2354-25G
B2354-5G
TCB2354-25GEA
133
USD
TCB2354-25G
TCB2354-5G
3-Bromo-p-toluidine ≥98.0%
3-Bromo-p-toluidine
CAS Number: 7745-91-7
MDL Number: MFCD00134176
Molecular Formula: C7H8BrN
Molecular Weight: 186.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 139
Flash Point (°C): 113
Freezing point (°C): 28
Other Product Names / Synonyms: 4-Amino-2-bromotoluene, 3-Bromo-p-toluidine
Formula:
C₇H₈BrN MW: 186.05 Boiling Pt: 139 °C Flash Pt: 113 °C |
MDL Number:
MFCD00134176 CAS Number: 7745-91-7 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...