1-Butylimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole

B2345-25G B2345-100G
TCB2345-25GEA 27 USD
TCB2345-25G TCB2345-100G
1-Butylimidazole ≥98.0% (by GC, titration analysis)
1-Butylimidazole

CAS Number: 4316-42-1
MDL Number: MFCD00042753
Molecular Formula: C7H12N2
Molecular Weight: 124.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 145
Specific Gravity (20/20): 0.95

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 235 °C
Density: 0.95 (20 °C)
Flash Pt: 145 °C
MDL Number: MFCD00042753
CAS Number: 4316-42-1
UN: 2690
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a4c0994583cd4a57df7898b7a9406c6f9824b1a&vid=5a4c0994583cd4a57df7898b7a9406c6f9824b1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR