4-Bromocrotonic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: trans-4-Bromo-2-butenoic Acid, (E)-4-Bromobut-2-enoic acid

B2298-1G B2298-5G
TCB2298-1GEA 208.2 USD
TCB2298-1G TCB2298-5G
4-Bromocrotonic acid ≥98.0% (by GC, titration analysis)
4-Bromocrotonic acid

CAS Number: 13991-36-1
MDL Number: MFCD00082701
Molecular Formula: C4H5BrO2
Molecular Weight: 164.99
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 74
Storage Temperature: 0-10°C


Other Product Names / Synonyms: trans-4-Bromo-2-butenoic Acid

Formula: C₄H₅BrO₂
MW: 164.99 g/mol
Melting Pt: 74 °C
MDL Number: MFCD00082701
CAS Number: 13991-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b5ac5720db787699a98abce139b72316416dea4&vid=0b5ac5720db787699a98abce139b72316416dea4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR