1-[Bis(trifluoromethanesulfonyl)methyl]-2,3,4,5,6-pentafluorobenzene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,3,4,5,6-Pentafluorophenylbis(trifluoromethanesulphonyl)methane, alpha,alpha-Bis(trifluoromethanesulphonyl)-2,3,4,5,6-pentafluorotoluene
CAS Number: 405074-81-9
MDL Number: MFCD04117910
Molecular Formula: C9HF11O4S2
Molecular Weight: 446.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 89
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: alpha,alpha-Bis(trifluoromethanesulfonyl)-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorophenylbis(trifluoromethanesulfonyl)methane
Formula:
C₉HF₁₁O₄S₂ MW: 446.21 g/mol Melting Pt: 89 °C |
MDL Number:
MFCD04117910 CAS Number: 405074-81-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...