1-(4-Bromophenyl)ethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromo-α-methylbenzylamine, (+/-)-1-(4-Bromophenyl)ethylamine, (±)-4-Bromo-α-methylbenzylamine

B2266-25G B2266-5G
TCB2266-25GEA 337 USD
TCB2266-25G TCB2266-5G
1-(4-Bromophenyl)ethylamine ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)ethylamine

CAS Number: 24358-62-1
MDL Number: MFCD00025548
Molecular Formula: C8H10BrN
Molecular Weight: 200.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.40


Other Product Names / Synonyms: 4-Bromo-alpha-methylbenzylamine

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Density: 1.40 (20 °C)
MDL Number: MFCD00025548
CAS Number: 24358-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22595e5c1b0b535339496af8c13e9f7157ecd560&vid=22595e5c1b0b535339496af8c13e9f7157ecd560&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR