4-tert-Butyl-α-chlorotoluene ≥89.0%
Supplier: TCI America
Synonyms:
p-tert-Butylbenzyl chloride, 4-tert-Butylbenzyl chloride, p-tert-Butyl-α-chlorotoluene, 1-(tert-Butyl)-4-(chloromethyl)benzene
B2235-25G
B2235-5G
TCB2235-25GEA
84.7
USD
TCB2235-25G
TCB2235-5G
4-tert-Butyl-α-chlorotoluene ≥89.0%
4-tert-Butyl-α-chlorotoluene
CAS Number: 19692-45-6
MDL Number: MFCD00000918
Molecular Formula: C11H15Cl
Molecular Weight: 182.69
Purity/Analysis Method: >89.0% (GC)
Form: Clear Liquid
Flash Point (°C): 94
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: 4-tert-Butyl-alpha-chlorotoluene
Formula:
C₁₁H₁₅Cl Density: 1.01 (20 °C) Flash Pt: 94 °C |
MDL Number:
MFCD00000918 CAS Number: 19692-45-6 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >89.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...