Methyl-3-(bromomethyl)benzoate ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Methyl 3-(bromomethyl)benzoate
B2224-25G
B2224-5G
TCB2224-25GEA
148.6
USD
TCB2224-25G
TCB2224-5G
Methyl-3-(bromomethyl)benzoate ≥97.0% (by GC)
Methyl 3-(bromomethyl)benzoate
CAS Number: 1129-28-8
MDL Number: MFCD00051437
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 169
Melting point (°C): 44
Other Product Names / Synonyms: 3-(Bromomethyl)benzoic Acid Methyl Ester, alpha-Bromo-m-toluic Acid Methyl Ester, Methyl alpha-Bromo-m-toluate
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 169 °C Melting Pt: 44 °C |
MDL Number:
MFCD00051437 CAS Number: 1129-28-8 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...