3-Bromobenzyl chloride ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Bromo-α-chlorotoluene, 1-(Chloromethyl)-3-bromobenzene, α-Chloro-3-bromotoluene, 1-Bromo-3-(chloromethyl)benzene

B2213-25G B2213-5G
TCB2213-25GEA 142 USD
TCB2213-25G TCB2213-5G
3-Bromobenzyl chloride ≥98.0% (by GC)
3-Bromobenzyl chloride

CAS Number: 932-77-4
MDL Number: MFCD00040865
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113


Other Product Names / Synonyms: 3-Bromo-alpha-chlorotoluene

Formula: C₇H₆BrCl
MW: 205.48 g/mol
Flash Pt: 113 °C
MDL Number: MFCD00040865
CAS Number: 932-77-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e4c00a87508d3c0d208237d3c71a888eca102eb&vid=4164caf962f37479b40e68154d4be008165e1530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR