1,4-Di(bromomethyl)-2,5-dimethylbenzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1,4-Bis(bromomethyl)-2,5-dimethylbenzene, 2,5-Bis(bromomethyl)-p-xylene, 2,5-Dimethyl-1,4-bis(bromomethyl)benzene
B2210-1G
B2210-5G
TCB2210-1GEA
126.6
USD
TCB2210-1G
TCB2210-5G
1,4-Di(bromomethyl)-2,5-dimethylbenzene ≥98.0% (by GC)
1,4-Di(bromomethyl)-2,5-dimethylbenzene
CAS Number: 35168-62-8
MDL Number: MFCD04038409
Molecular Formula: C10H12Br2
Molecular Weight: 292.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 163
Other Product Names / Synonyms: 2,5-Bis(bromomethyl)-p-xylene, 2,5-Dimethyl-1,4-bis(bromomethyl)benzene
Formula:
C₁₀H₁₂Br₂ MW: 292.01 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD04038409 CAS Number: 35168-62-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...