2-Bromo-6-nitrophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B2203-25G
B2203-5G
TCB2203-25GEA
176.8
USD
TCB2203-25G
TCB2203-5G
2-Bromo-6-nitrophenol ≥98.0% (by GC, titration analysis)
2-Bromo-6-nitrophenol
CAS Number: 13073-25-1
MDL Number: MFCD02683216
Molecular Formula: C6H4BrNO3
Molecular Weight: 218.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 68
Other Product Names / Synonyms: 6-Nitro-2-bromophenol
Formula:
C₆H₄BrNO₃ MW: 218.01 g/mol Melting Pt: 68 °C |
MDL Number:
MFCD02683216 CAS Number: 13073-25-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...