Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur ≥96.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 32133-82-7
MDL Number: MFCD00010662
Molecular Formula: C30H20F12O2S
Molecular Weight: 672.53
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 111
Storage Temperature: <0°C
Other Product Names / Synonyms: Martin Sulfurane
Formula:
C₃₀H₂₀F₁₂O₂S Melting Pt: 111 °C |
MDL Number:
MFCD00010662 CAS Number: 32133-82-7 |
Specification Test Results
Purity/Analysis Method | >96.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...