Biphenyl-4-sulfonyl chloride ≥98.0%

Supplier: TCI America

Synonyms: Biphenyl-4-sulphonyl Chloride

B2199-25G B2199-5G
TCB2199-25GEA 214 USD
TCB2199-25G TCB2199-5G
Biphenyl-4-sulfonyl chloride ≥98.0%
Biphenyl-4-sulphonyl chloride

CAS Number: 1623-93-4 MDL Number: MFCD00078658 Molecular Formula: C12H9ClO2S Molecular Weight: 252.71 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 114

Formula: C₁₂H₉ClO₂S
Melting Pt: 114 °C
MDL Number: MFCD00078658
CAS Number: 1623-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08b89cdaacff9ddf227b12c4e5bbc6610c993870&vid=08b89cdaacff9ddf227b12c4e5bbc6610c993870&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR