1-Butyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

B2194-100G B2194-25G B2194-5G
TCB2194-100GEA 156.58 USD
TCB2194-100G TCB2194-25G TCB2194-5G
1-Butyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
1-Butyl-3-methylimidazolium chloride

CAS Number: 79917-90-1
MDL Number: MFCD03095425
Molecular Formula: C8H15ClN2
Molecular Weight: 174.67
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 192

Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
Flash Pt: 192 °C
MDL Number: MFCD03095425
CAS Number: 79917-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3fee9e4cc7d9a12d5a4b03a1c69e225a2ae336a&vid=b3fee9e4cc7d9a12d5a4b03a1c69e225a2ae336a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR